Learn More
Novus Biologicals™ Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-89440
Product Code. 18211472
  • £328.00 / Pack of 2
Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
The Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Spezifikation
Spezifikation
Western Blot
2 vials
Universal Protein Lysate: Rat Normal Tissues
Normal
Rat
Rat Universal Protein Whole Tissue Lysate (Adult Normal)
Adult
Universal
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Rat
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
For Research Use Only