missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat (Wistar) Cryopreserved Hepatocytes, Plateable Male

Product Code. 13365664
Click to view available options
Quantity:
1 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13365664

Brand: Gibco™ WICP10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat (Wistar) Cryopreserved Hepatocytes, Plateable Male

These plateable cryopreserved male rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4–8 million

Get the Results You Need with GIBCO Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitro⁄in vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:

  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%
  • Attachment efficiency
  • Monolayer formation and integrity

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes

    Learn about our full line of hepatocytes,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Cryopreserved Hepatocytes stored in liquid nitrogen (LN2).
    Cell Type Hepatocytes
    Form Cryopreserved
    Gender Male
    Species Rat
    Donor Source Pooled
    No. of Cells 4-8 x 106
    Product Line Gibco
    Product Type Hepatocytes
    Quantity 1 mL

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.