Learn More
Reagecon™ Ratio Turbidity Standard
Brand:  Reagecon™ CRSR-20-100
 View more versions of this product
Product Code. 11788455
  • £108.00 / 100mL
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Ratio Turbidity Standard
±1%
NIST, US EPA Approved
Ratio Instruments
20 NTU
100 mL