missing translation for 'onlineSavingsMsg'
Learn More

RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17250-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847104

  • £130.00 / 20µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1CC1. the calculated molecular weight of RB1CC1 is 180 kDa, but the modified RB1CC1 is about 200 kDa.

The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

RB1CC1
Polyclonal
Unconjugated
RB1CC1
CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC
Rabbit
Antigen Affinity Chromatography
RUO
12421, 312927, 9821
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TDY2, Q9ESK9
Rb1cc1
RB1CC1 Fusion Protein Ag10508
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers