Learn More
Invitrogen™ eBioscience™ 1X RBC Lysis Buffer
Formulated for optimal lysis of erythrocytes in single-cell suspensions of mouse hematopoietic tissues such as spleen and human peripheral blood.
Brand: Invitrogen™ 00-4333-57
35.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.01000kg
Description
- Contains ammonium chloride, which lyses red cells with minimal effect on lymphocytes when used as instructed
- Nucleated red cells are not effectively lysed with ammonium chloride
Specifications
eBioscience™ | |
Liquid | |
200 mL |
200 mL | |
Lysis Buffer |
For Research Use Only. Not for use in diagnostic procedures.