Learn More
Thermo Scientific Chemicals RBC lysis buffer for human
For lysis of human red blood cells
£54.57 - £85.73
Specifications
Format | Liquid |
---|---|
Product Type | Cell Lysis Reagent |
For Use With (Application) | For lysis of human red blood cells. |
Content And Storage | 2° to 8°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465269
|
Thermo Scientific Chemicals
J62990.AK |
250mL |
£54.57
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15475269
|
Thermo Scientific Chemicals
J62990.AP |
500mL |
£85.73
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For lysis of human red blood cells and can be used to lyse reticulocytes, erythrocytes, and mature RBCs.
RBC Lysis Buffer for human is used in the isolation of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Liquid | |
Cell Lysis Reagent | |
For lysis of human red blood cells. | |
2° to 8°C |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.