Learn More
Thermo Scientific Chemicals RBC lysis buffer for mouse
For lysis of mouse red blood cells
£55.68 - £87.44
Specifications
Format | Liquid |
---|---|
Product Type | Cell Lysis Reagent |
For Use With (Application) | For lysis of mouse red blood cells |
Content And Storage | 2° to 8°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15474569
|
Thermo Scientific Chemicals
J62150.AK |
250mL |
£55.68
250mL |
In Stock
Log in to see stock. |
|||||
15484569
|
Thermo Scientific Chemicals
J62150.AP |
500mL |
£87.44
500mL |
In Stock
Log in to see stock. |
|||||
Description
RBC Lysis Buffer has been reported for use in flow cytometric analysis and cell culture. Red Blood Cell Lysing Buffer has been developed for use in hybridoma protocols to remove red blood cells from mouse splenocyte suspensions before fusion. It is also useful in systems where it may be desirable to remove red blood cells from cell suspensions, such as whole blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Liquid | |
Cell Lysis Reagent | |
For lysis of mouse red blood cells | |
2° to 8°C |
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.