missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ RBC and PLT Control Kit (CE/IVD)
To monitor methods for enumeration of residual leucocytes in leucoreduced platelet products.
Brand: BD Biosciences™ 341003
Additional Details : Weight : 0.05710kg
Description
- Used to monitor methods for enumeration of residual leucocytes in leucoreduced platelet products, including the dilution and staining process, method setup, and WBC enumeration.
- This product is CE marked to the IVD directive 98/79/EC and available in Europe for use on the BD FACSVia™ system and the BD FACSCalibur™ flow cytometer.
Specifications
CE marked to the IVD directive 98/79/EC | |
Monitor Enumeration | |
Flow Cytometer | |
Opened vials are stable for 30 days or 21 thermal cycles (uses), whichever comes first, when handled properly. A thermal cycle constitutes performing all steps once under Instructions for Use. In general, stability is limited by the number of times the vial is removed from the refrigerator, warmed, and mixed (defined as a use ); by the time from first use; and by the decrease in volume with each use. |
Store vials upright, tightly capped, at 2°C to 8°C when not in use. Unopened vials are stable until the expiration date indicated on each vial and on the Assay Values sheet when stored continuously at 2°C to 8°C. Protect from freezing, from temperatures above 30°C, and from prolonged time at room temperature (18°C to 30°C). | |
RBC/PLT Control | |
25 Tests |