missing translation for 'onlineSavingsMsg'
Learn More

RBM8A/Y14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen RBM8A/Y14
Concentration 0.22 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16863894
View Documents
Proteintech
14958-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16853894
View Documents
Proteintech
14958-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a protein with a conserved RNA-binding motif. The protein is found predomitly in the nucleus, although it is also present in the cytoplasm. It is preferentially associated with mRNAs produced by splicing, including both nuclear mRNAs and newly exported cytoplasmic mRNAs. It is thought that the protein remains associated with spliced mRNAs as a tag to indicate where introns had been present, thus coupling pre- and post-mRNA splicing events. Previously, it was thought that two genes encode this protein, RBM8A and RBM8B; it is now thought that the RBM8B locus is a pseudogene. Two alternative start codons result in two forms of the protein, and this gene also uses multiple polyadenylation sites.
Specifications

Specifications

RBM8A/Y14
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Rat, Human, Mouse
Q27W01, Q9CWZ3, Q9Y5S9
295284, 60365, 9939
RBM8A,Y14 Fusion Protein Ag6800
Primary
-20°C
RBM8A
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Binder of OVCA1 1, BOV 1, BOV 1A, BOV 1B, BOV 1C, MDS014, RBM8, RBM8A, RBM8B, Ribonucleoprotein RBM8A, RNA binding motif protein 8A, RNA binding protein 8A, RNA binding protein Y14, Y14, ZNRP, ZRNP1
Rbm8a
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications