missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14352-1-AP-150UL
This item is not returnable.
View return policy
Description
IRBP shuttles 11-cis and all trans retinoids between the retinol isomerase in the pigment epithelium and the visual pigments in the photoreceptor cells of the retina.Specifications
RBP3 | |
Polyclonal | |
Unconjugated | |
RBP3 | |
D10S64, D10S65, D10S66, IRBP, RBP3, RBPI, Retinol binding protein 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19661, 24711, 5949 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10745, P49194 | |
RBP3 | |
RBP3 Fusion Protein Ag5699 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |