missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ RC 2 Two-position Hot Plate

RC 2 Two-position Hot Plate with an aluminum top and temperature regulation.

Brand:  Velp Scientifica™ F20700620

Product Code. 30042335

  • £499.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Excellent Heating Performance for all lab applications

  • The RC 2 is equipped with two 135 mm diameter aluminum alloy top plates ensuring great temperature homogeneity and optimum heat transfer across the entire surface
  • Reliable for everyday laboratory applications such as boiling liquids, basic solutions, and media preparation as well as dissolving buffers and reagents
  • By turning the knobs of each independent position, it is possible to easily adjust the temperature of the plate up to 310°C
  • The RC 2 accepts up to 2 flasks of 15 liters each (H2O)

Easy-to-use, Safe and Designed to Last

  • An extremely intuitive control panel and its inclination have been carefully studied to optimize the user experience for all the lab operators and to offer great visibility
  • The RC 2 hotplate ensures a high level of safety and protection for laboratory operators
  • Internal electronic parts are protected by the insulating disk, which, together with the robust die-cast aluminum housing makes the RC 2 a durable and reliable hotplate
  • Features various safety functions, such as safety circuits and over-temperature protection, protect users in their daily operations
  • Two illuminated indicators always inform the operator when the heating is on
  • The icons will stay on when the plate's temperature is above 50°C, even if the instrument is switched off
Specifications

Specifications

Epoxy painted aluminum structure
Heating Plate
2
Round
3 Years
135 mm dia.
340 x 90 x 270 mm
Analog
EU/UK/AUS/CN
230 V
Aluminum Alloy
0°C to 300°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers