Learn More
Velp Scientifica™ RC 5 Hotplate
Single-position heating plates with aluminium top.
Brand: Velp Scientifica™
Additional Details : Weight : 1.40000kg
Description
The RC 5 is equipped with a 135 mm diameter aluminum alloy top plate ensuring great temperature homogeneity and optimum heat transfer across the entire surface.
You can rely on the RC 5 for everyday laboratory applications such as boiling liquids, basic solutions and media preparation, and dissolving buffers and reagents.
The RC 5 enables every laboratory to meet the highest safety needs: various safety functions, such as safety circuits, over-temperature protection and probe detection alarm, protect the user in daily operations. It is also possible to limit the maximum operating temperature.
Thanks to the insulation of the heating place and a robust die-cast aluminium housing, all the internal electronic parts are protected, enabling the instruments to work even in the most demanding laboratory environments.
The highly intuitive control panel and its inclination have been carefully studied to optimize the user experience for all the lab operators and to offer great visibility.
Specifications
EU-Style | |
160 x 270 x 85 mm | |
15 L | |
Hot Plate | |
CE | |
Round | |
Aluminum Alloy | |
135 mm dia. | |
1.4 kg | |
50/60 Hz |
None | |
Up to 15 L | |
1 | |
3 Years | |
230 V | |
Aluminum | |
Analog | |
50°C to 310°C | |
600 W | |
Over 50°C (overtemperature alarm over 330°C) |