missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ RD40 Plastic Filter AB2P3
Cost-effective plastic filter protects against hazardous gases, vapors, and particulates.
Brand: Honeywell™ 1788070
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- For use with full-face mask and half-mask equipped with an RD40 connector
- For protection against organic gases and vapors with a boiling point greater than 65°C and solid and liquid particles, inorganic gases and vapors
- Light and resistant plastic casing
- Suitable for use in metal-free zones and nuclear industry
- Performance requirements for gas and particles significantly above standard for excellent filtration efficiency over extensive period of time
- Canister made of high-grade charcoal, additionally treated for optimal gas absorption
- Specific canister structure for low breathing resistance and increased user comfort
- Low profile for unobstructed field of vision
- Multipurpose: can be used with existing respirators
- Safe storage between periods of use (plastic cover)
Specifications
EC PPE: 3, QA: ISO 9001/2000, EC Cert: 0070-236-079-11-03-0089, EC Attestation: 0070-236-079-11-03-0089, Declaration of conformity: APAVE-1788070 | |
Activated and impregnated charcoal without chromium, 106 mm OD, 300 gr | |
5/Pk. |
RD40 | |
ABS plastic with plastic cover |