missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RDH10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | RDH10 |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803394
|
Proteintech
14644-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16893384
|
Proteintech
14644-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RDH10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RDH10. This antibody recognizes human, mouse, rat antigen. The RDH10 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
RDH10 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
300 μg/mL | |
Polyclonal | |
Rabbit | |
157506 | |
Primary |