Learn More
RDM1 Mouse anti-Human, Unconjugated, Clone: 1C10B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66941-1-IG-150UL
 View more versions of this product
Product Code. 16819255
  • £349.00 / 150µL
Explore more special offers
Description
Description
The RDM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RDM1. This antibody recognizes human, mouse, rat antigen. The RDM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
RDM1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1C10B4
Unconjugated
150 μL
201299
IgG2a