missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ rDNase Set
Click to view available options
Quantity:
1 Set
Unit Size:
50 sets
Descripción
The rDNase Set is designed for on-column DNA removal using any NucleoSpin RNA Kit. (The number of preps depends on the exact kit.) The rDNase Set is also suitable for digestion of contaminating DNA within pre-purified RNA preparations in solution (e.g. phenol-based RNA preparations).
- On-column DNA digestion—Time-saving DNA removal during RNA preparation
- DNA digestion in solution —Allows most efficient DNA removal for sensitive applications
- Lyophilized rDNase from bovine pancreas
- Recombinantly produced in Pichia pastoris without using any animal cells
- No RNase activity detectable
- RNA integrity (RIN) is not affected by rDNase treatment
- Separate reaction buffer optimized for best rDNase performance
Especificaciones
Especificaciones
| Components | Lyophilized enzyme: reconstitude by adding 550μL RNase-free water in supplied vial |
| For Use With (Application) | RNA purification—Removal of contaminating DNA |
| For Use With (Equipment) | NucleoSpin™ RNA kits |
| Packaging | Vial with lid |
| Content And Storage | Store Lyophilized rDNase at +4 °C On Arrival Store at 4°C for up to 1 year Store Reconstituted rDNase at −20°C for up to 6 months |
| Source | Bovine pancreas, recombinantly produced in Pichia pastoris without using any animal cells or other material derived from animals |
| Quantity | 1 Set |
| Product Type | rDNase Set |
Seguridad y manipulación
Palabra de advertencia
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido