Learn More
Thermo Scientific™ Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Use these triangular stir bars for faster reaction times and smooth mixing in vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System.
£150.96 - £251.81
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
11597450
|
Thermo Scientific™
TS-16010 |
0.3 and 1mL Reacti-Vials | 6 Pack |
£150.96
Pack of 6 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
11587450
|
Thermo Scientific™
TS-16000 |
3, 5, and 10 mL Reacti-Vials | 6 Pack |
£251.81
Pack of 6 |
In Stock
Log in to see stock. |
||||
Description
Use these triangular stir bars for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminum Blocks.
Specifications
6 Pack |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.