Learn More
Thermo Scientific™ Reacti-Therm™ Thermometers
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
Brand:  Thermo Scientific™ TS-18915
 View more versions of this product
Product Code. 11547510
  • £138.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermometer
0°C to 200°C
Dry Block Heaters and Other Instruments
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.