missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Reaction Buffer for rDNase
Included in the rDNase Set (ref 740963) for efficient on-column removal of contaminating DNA contamination or DNA digestion in solution during isolation of pure RNA using compatible kits.
Brand: Macherey-Nagel™ 740834.60
This item is not returnable.
View return policy
Description
Compatible kits include:
- NucleoSpin™ RNA
- NucleoSpin™ miRNA
- NucleoSpin™ miRNA Plasma
- NucleoSpin™ RNA/Protein
- NucleoSpin™ TriPrep
- NucleoSpin™ RNA Blood
- NucleoSpin™ RNA Plant
- NucleoSpin™ totalRNA FFPE
- NucleoSpin™ RNA Clean-up
- NucleoMag™ DNA/RNA Water
Specifications
Colorless | |
6.5 - 7.5 | |
Liquid | |
Odorless |
1 - <2 % | |
Reaction buffer for rDNase | |
Isolation of pure RNA, removal of contaminating DNA | |
60 mL |