missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Reaction Vessels, Cylindrical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£605.00 - £624.00
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11552204
|
Lenz Laborglasintrumente
06007128 |
120 mm |
£605.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
11562204
|
Lenz Laborglasintrumente
06007137 |
185 mm |
£611.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
11572204
|
Lenz Laborglasintrumente
06007149 |
250 mm |
£624.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- With graduation
- With laboratory flange LF
- With groove
- With round bottom
- Made of DURAN™ tubing
- Other sizes on request
Specifications
60 mm |