missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Flat Flange Reaction Vessels, PTFE

Use these reaction vessels for a variety of distillation applications. BOLA™ PTFE Flat Flange Reaction Vessels feature, a round bottom, thick wall and smooth interior surface.

Brand:  Bohlender™ B281-09

Product Code. 10767682

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Can be heated by a thermostat or an electric heating mantle
  • Temperature resistance: -200°C to +200°C
  • Universal chemical resistance
  • SUitable vacuum
Specifications

Specifications

270mm
2,000mL
Flange: 100NW
PTFE
140mm
-200°C to +200°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bohlender™ BOLA™ Flat Flange Reaction Vessels, PTFE > Flat Flange Reaction Vessels, PTFE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.