Learn More
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
£444.00 - £1325.00
Specifications
Content And Storage Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Description illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
For Use With (Application) Genomic DNA Amplification
Size > 10 kb
Test Time 4 h
View More Specs
Products 2
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
12987160
View Documents
Cytiva
25-6603-24
1 x 2.6mL 2X Denaturation Buffer, 3 x 8-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps 24
£444.00
Each
12997160
View Documents
Cytiva
25-6603-96
1 x 2.6mL 2X Denaturation Buffer, 3 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps 96
£1325.00
Each
Description
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
Specifications
Specifications
Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons
Genomic DNA Amplification
4 h
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
> 10 kb
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit