missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Ready-To-Use Lysing Bead Tubes for Bead Mill Homogenizers
Protect the integrity of your samples - tubes, grinding beads and balls are treated and tested to be DNase and RNase-free.
£323.00
Specifications
Product Type | Lysing Bead Tubes |
---|---|
Capacity (Metric) | 2 mL |
Material | Polyethylene |
Bottom Type | Skirted Tube |
For Use With (Equipment) | Homogenizers |
Product Code | Brand | Closure Color | Sample Type | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Sample Type | Particle Size | Price | Quantity & Availability | |||
17949530
|
OHAUS™
30391409 |
Red | Animal Tissue | 2.8 mm |
£323.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Convenient wet or dry grinding
- Seven distinct media types for plant, soft sample, fungi, yeast, bacteria or environmental sample processing
- Threaded caps are color coded for easy identification
- Applications:
- DNA, RNA and Protein Analysis (from mammalian tissue, plant, soil, seed, fungi, bacteria, virus and other cells), Cell Lysing, PCR, Gene Expression
- Design Features:
- Tubes and grinding media are DNase/RNase-free
Specifications
Lysing Bead Tubes | |
Polyethylene | |
Homogenizers |
2 mL | |
Skirted Tube | |
100 Pk. |