missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ 2.5% Normal Horse Serum (1X)

Product Code. 17112514
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 17112514

Brand: Invitrogen™ R37625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments

  • ReadyProbes 2.5% Normal Horse Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments, usually when a horse or donkey secondary antibody is used.
  • Incubating cells/tissue with diluted serum from the same or similar species as the host of the secondary antibody typically reduces non-specific binding.
  • Simply cover the tissue section or cells with a few drops of ReadyProbes 2.5% Normal Horse Serum and incubate for 30–60 minutes at room temperature.
  • Ready Probes 2.5% Normal Horse Serum can also be used to dilute primary and secondary antibodies.
  • Ready Probes 2.5% Normal Horse Serum is provided as a ready-to-use reagent with no further dilution or preparation required.
  • The serum is collected from healthy adult animals and the heat-treated, centrifuged, and filtered. It is then checked for cross reactivity using an appropriate immunoassay.
  • The serum is diluted to 2.5% (v/v) in buffer, with 0.08% sodium azide added as a preservative.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Provided as 50-mL solution at ready-to-use (1X) concentration in a clear plastic dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Species Horse
Form Liquid
Product Type Horse Serum
Quantity 50 mL
Product Line ReadyProbes
Reagent Type Blocking Reagent
Shipping Condition Room Temperature or Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.