missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X)

Inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissue sections, frozen tissue sections, and cell preparations

Brand:  Invitrogen™ R37629

104.88 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 17163773

  • £138.00 / 100mL
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissue sections, frozen tissue sections, and cell preparations.
  • It is provided in a ready-to-use format—just add a few drops and incubate for 10–15 minutes.
  • Endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activities become problematic when alkaline phosphatase (AP)- or horseradish peroxidase (HRP)- based detection systems are used.
  • ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits these endogenous activities, for reduced non-specific signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures. This blocker is used before any standard, non-specific blocking solutions (e.g., diluted serum).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocking Solution
ReadyProbes™
Provided as 100-mL solution at ready-to-use (1X) concentration in a white dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

100 mL
Liquid
Approved for shipment at Room Temperature or on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-31

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X) > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.