Learn More
Invitrogen™ ReadyProbes™ Tissue Autofluorescence Quenching Kit
Minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin
Brand:  Invitrogen™ R37630
122.36 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Product Code. 17162534
  • £165.00 / 1 set
Edge
Explore more special offers
Description
Description
  • The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin.
  • If the target signal is low enough, then this background signal can interfere with the interpretation of immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence(IF), and in situ hybridization (ISH) assays.
  • The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes most kinds of autofluorescence, with the exception of autofluorescence from lipofuschin granules.
  • The ReadyProbes Tissue Autofluorescence Quenching Kit contains three components that are combined just before use. Combine components A, B, and C in a 1:1:1 ratio.
  • Mix components A and B together before adding component C. The resulting solution is then applied to the tissue for 2–5 minutes at room temperature.
  • This is done after the blocking, primary antibody, and secondary antibody steps, but before application of any counterstains like nuclear stains.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Approved for shipment at Room Temperature or on Wet Ice
Blocking Reagent
A kit containing 5 mL each ready-to-use (1X) solutions of reagent A, B, and C in three clear HDPE bottles.
  • Store at 2–8°C.
  • When stored as directed, product can be used for 3 months after receipt.One kit is sufficient to tre
    ReadyProbes™
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Invitrogen™ ReadyProbes™ Tissue Autofluorescence Quenching Kit > 1 kit