missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck

Catalog Number 15221819
125.58 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.


Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Classic narrow-neck, glass-stoppered reagent bottles in amber soda lime glass.

A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!

  • UV protection due to external colouration
  • All glass components
  • Tight closure due to standard ground joint
Capacity (Metric) 50 mL
For Use With (Application) Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules
Includes With standard ground glass flat-headed stopper
Material Soda Lime Glass
Quantity 10/Pk.
Autoclavable Not Autoclavable
Diameter (Metric) 42 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'