missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck
Classic narrow-neck, glass-stoppered reagent soda lime glass bottles.
£99.80 - £265.00
Specifications
For Use With (Application) | Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules |
---|---|
Includes | With standard ground glass flat-headed stopper |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15261809
|
DWK Life Sciences
231655405 |
1000 mL |
£189.00
Pack of 10 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
15241809
|
DWK Life Sciences
231652408 |
100 mL |
£99.80
Pack of 10 |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
11830700
|
DWK Life Sciences
231653601 |
250 mL |
£123.00
Pack of 10 |
Estimated Shipment: 29-10-2025
Log in to see stock. |
|||||
15251809
|
DWK Life Sciences
231654409 |
500 mL |
£265.00
Pack of 10 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- All glass components
- Tight closure due to standard ground joint
Specifications
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules | |
Soda Lime Glass | |
Not Autoclavable |
With standard ground glass flat-headed stopper | |
10/Pk. |