Learn More
Thermo Scientific™ Reagent Kits for Eutech™ Colorimeters
Reagent Kits used with Eutech Colorimeters
Brand:  Thermo Scientific™ 94X377005
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11519304
  • £31.49 / Pack of 100
In Stock
to see stock.
Description
Description
The Reagent kits are used with Eutech™ Colorimeters C401, C301, C201, C105, C104, C103, C102 and C101.
Eutech™’s latest range of laboratory grade microprocessor-based portable waterproof colorimeters measure Chlorine (Free and Total), Cyanuric Acid, pH, Chlorine Dioxide, Bromine and Ozone with quick, accurate and repeatable results. The meters are easy to use because of their simple, intuitive and direct user interface. Guiding messages and self-diagnostics are displayed on a large custom-made liquid crystal display (LCD). Each meter comes with reagents and sample vials in a rugged carrying case for swift testing-on-the-go. This family of colorimeters is designed for reliable and convenient water analysis in swimming pools, spas, public utilities, industrial wastewater plants, municipal water, treated water, water conditioning systems and paper and pulp mills. Select from our wide range of models, designed to meet your application needs.
Colorimeter Features:
  • Advanced Powermanagement
    The meter is capable of measuring more than 3,000 tests using a single set of 4 'AAA' alkaline batteries. Auto power-off activates after 10 minutes of non-use to conserve battery life.
  • Fuss-freemeter Operation
    The meter is designed for easy operation. It allows one-time blanking for all parameters and ranges when measuring the same sample. The blanking value is stored in memory even if batteries are replaced. Furthermore, no calibration is needed for Chlorine and Cyanuric Acid. Most importantly, the meter is maintenance-free.
  • Large Easy-to-read Display
    The large screen features 4-digit 14-segment customized liquid crystal display (LCD) with mode indicators to guide meter use.
  • IP67 waterproof and lightweight
    The lightweight meter has a waterproof IP67-rated housing. It even floats when accidentally dropped in water.
Specifications
Specifications
Glycine (DPD) Reagent Kit, pack of 100 sachets (Used as Chlorine Dioxide Reagent)