missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reagent Reservoir Chiller

Product Code. 16350412 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
100 mL
50 and 55 mL
Color:
Blue
Purple
Purple & Blue
Unit Size:
Pack of 2

Product Code. 16350412

Only null left
Add to basket
Edge
Add to basket

The Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.

  • Reagents are kept at less than 5°C (41°F) for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Stackable, save space when storing in the freezer or on the bench top
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Made from sonic welded, engineered polypropylene that ensures a leak free design
  • Not intended for autoclave, do not use with liquid nitrogen
  • Reusable
  • Two-year warranty

Specifications

Product Type Reservoir Chiller
Autoclavable Autoclavable
Color Purple & Blue
Description Reagent Reservoir Chiller
Device Holds Reagent Reservoirs
Material Polypropylene
Volume (Metric) 50 and 55 mL
For Use With (Application) Reagent
For Use With (Equipment) Reagent Reservoirs
Includes Chiller
Model 120732F
Quantity 1
Shape Rectangular
Warranty 2 Years
Dimensions (L x W) 5.6 x 2.9 in.
Dimensions (L x W x H) 5.6 x 2.9 x 1.75 in.
Length (English) 5.6 in.
Length (Metric) 14.4 cm
Width (English) 5.70 in.
Width (Metric) 14.30 cm
Height (English) 1.75 in.
Height (Metric) 4.5 cm
Weight (English) 0.31 lb.
Weight (Metric) 0.14 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.