Learn More
Fisherbrand™ Reagent Reservoir Chiller
The Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.
Brand: Fisherbrand™
14.18 GBP valid until 2024-08-31
Use promo code "21641" to get your promotional price.
Additional Details : Weight : 0.18000kg
Description
- Reagents are kept at less than 5°C (41°F) for approximately 3 hours
- Ice-free solution eliminates the risk of contamination from messy ice baths
- Stackable, save space when storing in the freezer or on the bench top
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
- Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
- Made from sonic welded, engineered polypropylene that ensures a leak free design
- Not intended for autoclave, do not use with liquid nitrogen
- Reusable
- Two-year warranty
Specifications
Autoclavable | |
Reagent Reservoir Chiller | |
Polypropylene | |
Reagent | |
Chiller | |
1 | |
2 Years | |
6 x 3.5 x 1.75 in. | |
15 cm | |
14.30 cm | |
4.3 cm | |
0.14 kg |
Blue | |
Reagent Reservoirs | |
100 mL | |
Reagent Reservoirs | |
120734F | |
Rectangular | |
6 x 3.5 in. | |
6 in. | |
5.70 in. | |
1.75 in. | |
0.31 lb. |