missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene
GREENER_CHOICEBIOPHARMA

Product Code. 16324862 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
Packaging:
1/bag, 100 bags
1/bag, 50 bags
5/bag, 20 bags
5/bag, 40 bags
Dimensions (L x W x H):
2.8 x 0.8 x 0.7 in
4 x 1 x 0.8 in
4 x 1.1 x 1.2 in
4.3 x 1.9 x 1.3 in
Unit Size:
Pack of 100
Pack of 200
Pack of 50

Product Code. 16324862

Brand: Heathrow Scientific™ 120647

Only null left
Add to basket
Edge
Add to basket

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures

  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
  • Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Quality certificate available on request
  • Disposable
  • Design Patent Pending

Specifications

Capacity (English) 3.38 oz
Certifications/Compliance Dnase, Rnase & Pyrogen Free
Coating None
Color White
Description 5 wrap
Capacity (Metric) 100 mL
Dimensions (L x W x H) 4.3 x 1.9 x 1.3 in
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) Reagent
Height (English) 1.3 in
Height (Metric) 3.2 cm
Includes Reservoir
Length (English) 4.3 in
Length (Metric) 10.9 cm
Max. Temperature (English) 104°F Operating
Max. Temperature (Metric) 40°C Operating
No. per Case 1200
No. per Pack 200
Packaging 5/bag, 40 bags
Quantity 200
Volume (Metric) Well 100 mL
Width (English) 1.9 in
Width (Metric) 4.8 cm
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Conical
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.