missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene

Shop All Thermo Scientific CO₂ Incubators Products


Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
Packaging:
1/bag, 100 bags
1/bag, 50 bags
5/bag, 20 bags
5/bag, 40 bags
Dimensions (L x W x H):
2.8 x 0.8 x 0.7 in
4 x 1 x 0.8 in
4 x 1.1 x 1.2 in
4.3 x 1.9 x 1.3 in
Unit Size:
Pack of 100
Pack of 200
Pack of 50
Description
- Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
- Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Quality certificate available on request
- Disposable
- Design Patent Pending
Specifications
Specifications
| Capacity (English) | .85 oz |
| Certifications/Compliance | Dnase, Rnase & Pyrogen Free |
| Coating | None |
| Color | White |
| Description | 5 wrap |
| Capacity (Metric) | 25 mL |
| Dimensions (L x W x H) | 4 x 1 x 0.8 in |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction