missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Reagent Reservoir, Sterile Polystyrene
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures
Brand: Heathrow Scientific™ 120643
127.29 GBP valid until 2024-12-31
Use promo code "22009" to get your promotional price.
Description
- Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents
- Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Quality certificate available on request
- Disposable
- Design Patent Pending
Specifications
1.69 oz | |
None | |
5 wrap | |
4 x 1.1 x 1.2 in | |
Sterile | |
Reagent | |
3 cm | |
4 in | |
104°F Operating | |
2000 | |
5/bag, 40 bags | |
50 mL | |
2.8 cm | |
1 | |
Conical |
Dnase, Rnase & Pyrogen Free | |
White | |
50 mL | |
Polystyrene | |
Reagent Reservoir | |
1.2 in | |
Reservoir | |
10.2 cm | |
40°C Operating | |
200 | |
200 | |
1.1 in | |
Not Autoclavable | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction