Learn More
Thermo Scientific™ Reagent Reservoirs
Choose these reagent reservoirs, designed to provide a variety of solutions to suit all of your pipetting procedures.
Brand: Thermo Scientific™ 95128093
169.37 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : Weight : 1.35000kg
Description
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Features:
- Trough within a trough maximizes the amount of liquid accessible to pipette tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
- 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
Specifications
White | |
Polystyrene | |
Reagent Reservoir |
25 mL | |
Non-sterile | |
Bulk |