missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set
Premeasured powder reagents in individual packages for determination of Ammonia Nitrogen. Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set include Ammonia Cyanurate and Ammonia Salicylate reagents that can be purchased together or used separately.
£71.00 - £114.00
Specifications
Analytes | Ammonia Nitrogen |
---|---|
Color | White |
Format | Individual Packet |
For Use With (Application) | Ammonia Nitrogen determination |
For Use With (Equipment) | All except DR/4000, DR/3000, DR/2000, DR 900 |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11926878
|
Hach™
2653199 |
Ammonia Cyanurate Reagent |
£71.00
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10472125
|
Hach™
2653299 |
Ammonia Salicylate Reagent |
£114.00
Pack of 100 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
Premeasured powder reagents in individual packages for determination of Ammonia Nitrogen. Hach™ Reagents for Nitrogen-Ammonia Powder Pillow Set include Ammonia Cyanurate and Ammonia Salicylate reagents that can be purchased together or used separately.
- Ready-to-use, premeasured powder reagents in individual sealed packets
- For Ammonia Nitrogen determination by the Salicylate Method (Hach Method 55)
- Ammonia Cyanurate and Ammonia Salicylate powder pillows for 100 tests
- Both sets are contained in the Hach Nitrogen-Ammonia Powder Pillow Reagent Set (man. no. 2668000)
Specifications
Ammonia Nitrogen | |
Individual Packet | |
All except DR/4000, DR/3000, DR/2000, DR 900 | |
100 Pack | |
10 mL | |
Reagent |
White | |
Ammonia Nitrogen determination | |
100 | |
0.01 to 0.50mg/L NH3-N | |
Salicylate |