missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Real Time Electrophoresis System, with Power Supply
Run DNA gels, view separations as they occur, and capture gel images using a smart phone
£1444.03
Specifications
Buffer | 0.5 x or 1 x TAE or TBE |
---|---|
Certifications/Compliance | CE |
Color | Gray and clear |
Detection Method | Smart phone |
For Use With (Equipment) | Standard Electrophoresis Power Supply |
Product Code | Brand | Includes | Plug Type | Voltage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Plug Type | Voltage | Price | Quantity & Availability | |||
17813388
|
Fisherbrand™
|
Gel tank, Blue LED illuminator, Filter cover, Gel casting set, Smart phone imaging enclosure, 230 to 115 V transformer | Universal | 220/240 V |
£1444.03
Each |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Tool for small DNA gel runs
- Power supply has selectable voltage of 100 V, 50 V, or 35 V and timer
- Compatible with SYBR, Gel green, SmartGlow, and other green fluorescent DNA stains
- Imaging enclosure and photo filter allows gel imaging with any smart phone camera
- Includes:
- Gel running tank
- Direct connect power supply
- Amber filter lid
- Small blue LED transilluminator
- Gel casting equipment (for casting your own gels)
- Stand
- Trays
- Combs
Specifications
0.5 x or 1 x TAE or TBE | |
Gray and clear | |
Standard Electrophoresis Power Supply | |
DNA | |
10.5 x 10 cm, 10.5 x 6 cm | |
Agarose | |
Use with SmartGlow or SYBR green stain | |
Electrophoresis | |
12 or 22 | |
10 to 20 μL | |
12 or 22 | |
230 mL | |
10 to 20 μL | |
2 years | |
7.5 x 8.25 x 5 in. | |
7.5 in. | |
21 in. | |
10.5 cm | |
3 mm, 5.65 mm | |
14 cm | |
0.75 kg |
CE | |
Smart phone | |
Make gels using included casting set | |
Digital LED timer | |
0.5 to 6% | |
Uses minimal consumables, no waste associated with precast gels | |
Molded polycarbonate | |
1 | |
1 Ea. | |
Nucleic acid | |
1 mm, 2 mm | |
230 mL | |
10 to 20 μL | |
7.5 x 8.25 in. | |
7.5 in. | |
8.25 in. | |
10 cm | |
3 mm, 5.65 mm | |
5 in. | |
1.65 lb. |