Learn More
Heidolph™ Reax 20/12
Resilient shaker that can hold up to 12 bottles.
Brand: Heidolph™ 541-20012-06
Additional Details : Weight : 32.20000kg
Description
- The Reax 20 is an ideal shaker for processing and storing samples that tend to sediment. The fields of application range from environmental technology to paint processing and quality assurance in the materials industry.
- The maximum permissible weight is 30 kg, so that samples with a high solid content can also be properly mixed. Since this device sets great forces in motion, it is equipped with an emergency stop switch for your safety.
- This overhead shaker can be ordered with a capacity of 4, 8 or 12 bottles. Also suitable for mixing cylinders or wide-neck bottles.
Specifications
230 V | |
UK Plug |