missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 2 subunit B Protein
A DNA sequence encoding the Enterobacteria phage 933W (Bacteriophage 933W) Shiga-like toxin 2 subunit B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7305-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_049501.1 | |
Shiga-like toxin 2 subunit B Protein | |
Research Use Only | |
stxB2 | |
Recombinant Protein | |
E. coli | |
ADCAKGKIEF SKYNEDDTFT VKVDGKEYWT SRWNLQPLLQ SAQLTGMTVT IKSSTCESGS GFAEVQFNND | |
Greater than 94.5% as determined by SDS-PAGE. |
1262010 | |
500 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |