missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 2 subunit B Protein
A DNA sequence encoding the Enterobacteria phage 933W (Bacteriophage 933W) Shiga-like toxin 2 subunit B, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_049501.1 |
---|---|
Gene ID (Entrez) | 1262010 |
Name | Shiga-like toxin 2 subunit B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058100
|
enQuireBio™
QP7305-ec-10ug |
10 μg | |||||||
16098100
|
enQuireBio™
QP7305-ec-50ug |
50 μg | |||||||
16048100
|
enQuireBio™
QP7305-ec-100ug |
100 μg |
£715.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16078100
|
enQuireBio™
QP7305-ec-200ug |
200 μg | |||||||
16088100
|
enQuireBio™
QP7305-ec-500ug |
500 μg | |||||||
16068100
|
enQuireBio™
QP7305-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_049501.1 | |
Shiga-like toxin 2 subunit B Protein | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1262010 | |
Research Use Only | |
stxB2 | |
Recombinant Protein | |
E. coli | |
ADCAKGKIEF SKYNEDDTFT VKVDGKEYWT SRWNLQPLLQ SAQLTGMTVT IKSSTCESGS GFAEVQFNND | |
Greater than 94.5% as determined by SDS-PAGE. |