missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Tail virion protein G9P Protein
A DNA sequence encoding the Enterobacteria phage M13 (Bacteriophage M13) Tail virion protein G9P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6944-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_510889.1 | |
Tail virion protein G9P Protein | |
Research Use Only | |
IX | |
Recombinant Protein | |
E. coli | |
MSVLVYSFAS FVLGWCLRSG ITYFTRLMET SS | |
Greater than 90% as determined by SDS-PAGE. |
927332 | |
50 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |