missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Tail virion protein G9P Protein

A DNA sequence encoding the Enterobacteria phage M13 (Bacteriophage M13) Tail virion protein G9P, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6944-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076810

  • £2725.00 / 1mg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_510889.1
Tail virion protein G9P Protein
Research Use Only
IX
Recombinant Protein
E. coli
MSVLVYSFAS FVLGWCLRSG ITYFTRLMET SS
Greater than 90% as determined by SDS-PAGE.
927332
1 mg
Not determined.
Full Length
Bacteriophage
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers