missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine FGF 2 Protein
A cDNA sequence encoding the FGF 2 was constructed and used to recombinantly synthesize the protein.
£136.00 - £320.00
Specifications
Gene ID (Entrez) | 281161 |
---|---|
Name | FGF 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | FGF2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974368
|
enQuireBio™
QP10599-2ug |
2 μg |
£136.00
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15954368
|
enQuireBio™
QP10599-10ug |
10 μg |
£320.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
281161 | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
Bovine | |
MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGPKTG PGQKAILFLP MSAKS | |
Greater than 95.0% as determined by SDS-PAGE. |
FGF 2 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The recommended concentration in responsive cells is 0.1 to 2ng/ml. | |
Bovine | |
Untagged | |
The FGF-basic Bovine was lyophilized from a concentrated sterile solution containing 50mM Na2HPO4, pH 7.5 and 0.5% HSA. |