missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine L2HGDH, mitochondrial Protein

A DNA sequence encoding the Bos taurus (Bovine) L2HGDH, mitochondrial, was expressed in the hosts and tags indicated.

£281.57 - £2635.00

Specifications

Accession Number NP_001094560.1
Gene ID (Entrez) 514230
Name L2HGDH, mitochondrial Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16082180
View Documents
enQuireBio™
QP6285-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16022190
View Documents
enQuireBio™
QP6285-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16072180
View Documents
enQuireBio™
QP6285-ec-100ug
100 μg
£762.00
100µg
Please to purchase this item. Need a web account? Register with us today!
16002190
View Documents
enQuireBio™
QP6285-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16012190
View Documents
enQuireBio™
QP6285-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16092180
View Documents
enQuireBio™
QP6285-ec-1mg
1 mg
£2635.00
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001094560.1
L2HGDH, mitochondrial Protein
Not determined.
Full Length
Bovine
Tris-based buffer, 50% glycerol
514230
Research Use Only
L2HGDH
Recombinant Protein
IIGGGIVGLA SARALILRHP ALSIGVLEKE KNLAVHQTGH NSGVIHSGIY YKPESLKAKL CVQGAALIYE YCNQKGISYK QCGKLIVAVE QEEIPRLQAL YERGLQNGVQ GLRLIQQEDI KKKEPYCRGL MAIDCPYTGI VDYRQVAFSF AKDFQEAGGS VLTNFEVEDI EMARESPSRS KDGMKYPIVI RNTKGEEVRC QYVVTCAGLY SDRISELSGC NPNPRIVPFR GDYLVLKPEK RYLVKGNIYP VPDSRFPFLG VHFTPRMDGN IWLGPNAILA FKREGYRPFD FSARDIMDII IKSGLIKLVF QNFSYGVNEM YKACFLSATV KHLQKFIPEI TISDVLRGPA GVRAQALDRD GNLIEDFVFD GGVGDIGNRI LHVRNAPSPA ATSSLAISGM IADEVQQRFK L
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Bovine L2HGDH, mitochondrial Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.