missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine p85a Protein

A cDNA sequence encoding the p85a was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12950-10ug

 View more versions of this product

Product Code. 15942049

  • £439.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

p85a Protein
Research Use Only
Recombinant Protein
Leishmania species
MSAEGYQYRA LYDYKKEREE DIDLHLGDIL TVNKGSLVAL GFSDGQEAKP EEIGWLNGYN ETTGERGDFP GTYVEYIGRK KISPPTPKPR PPRPLPVAPG PSKTEADSEQ QASTLPDLAE QFAPPDVAPP LLIKLVEAIE KKGLECSTLY RTQSSSNPAE LRQLLDCDTA SLDLEMFDVH VLADAFKRYL LDLPNPVIPV AVSSELISLA PEVQSSEEYI QLLKKLIRSP SIPHQYWLTL QYLLKHFFKL SQTSSKNLLN ARVLSELFSP LLFRFPAASS ENTEHLIKII EILISTEWNE RQPAPALPPK PPKPTTVANN GMNNNMSLQD AEWYWGDISR EEVNEKLRDT ADGTFLVRDA STKMHGDYTL TLRKGGNNKL IKIFHRDGKY GFSDPLTFNS VVELINHYRN ESLAQYNPKL DVKLLYPVSK YQQDQVVKED NIEAVGKKLH EYNTQFQEKS REYDRLYEDY TRTSQEIQMK RTAIEAFNET IKIFEEQCQT QERYSKEYIE KFKREGNETE IQRIMHNYEK LKSRISEIVD SRRRLEEDLK KQAAEYREID KRMNSIKPDL IQLRKTRDQY LMWLTQKGVR QKKLNEWLGN ENTEDQYSLV EDDEDLPHHD EKTWNVGSSN RNKAENLLRG KRDGTFLVRE SSKQGCYACS VVVDGEVKHC VINKTATGYG FAEPYNLYSS LKELVLHYQH TSLVQHNDSL NVTLAYPVYA QQRR
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Supplied as a 1 mg/ml solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers