missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine p85a Protein
A cDNA sequence encoding the p85a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12950-2ug
This item is not returnable.
View return policy
Specifications
p85a Protein | |
Research Use Only | |
Recombinant Protein | |
Leishmania species | |
MSAEGYQYRA LYDYKKEREE DIDLHLGDIL TVNKGSLVAL GFSDGQEAKP EEIGWLNGYN ETTGERGDFP GTYVEYIGRK KISPPTPKPR PPRPLPVAPG PSKTEADSEQ QASTLPDLAE QFAPPDVAPP LLIKLVEAIE KKGLECSTLY RTQSSSNPAE LRQLLDCDTA SLDLEMFDVH VLADAFKRYL LDLPNPVIPV AVSSELISLA PEVQSSEEYI QLLKKLIRSP SIPHQYWLTL QYLLKHFFKL SQTSSKNLLN ARVLSELFSP LLFRFPAASS ENTEHLIKII EILISTEWNE RQPAPALPPK PPKPTTVANN GMNNNMSLQD AEWYWGDISR EEVNEKLRDT ADGTFLVRDA STKMHGDYTL TLRKGGNNKL IKIFHRDGKY GFSDPLTFNS VVELINHYRN ESLAQYNPKL DVKLLYPVSK YQQDQVVKED NIEAVGKKLH EYNTQFQEKS REYDRLYEDY TRTSQEIQMK RTAIEAFNET IKIFEEQCQT QERYSKEYIE KFKREGNETE IQRIMHNYEK LKSRISEIVD SRRRLEEDLK KQAAEYREID KRMNSIKPDL IQLRKTRDQY LMWLTQKGVR QKKLNEWLGN ENTEDQYSLV EDDEDLPHHD EKTWNVGSSN RNKAENLLRG KRDGTFLVRE SSKQGCYACS VVVDGEVKHC VINKTATGYG FAEPYNLYSS LKELVLHYQH TSLVQHNDSL NVTLAYPVYA QQRR | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Supplied as a 1 mg/ml solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol. |