missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Serpin A3-2 Protein
A DNA sequence encoding the Bos taurus (Bovine) Serpin A3-2, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Accession Number | NP_001139773.1 |
---|---|
Gene ID (Entrez) | 100272170 |
Name | Serpin A3-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058601
|
enQuireBio™
QP9699-ye-10ug |
10 μg | |||||||
16098601
|
enQuireBio™
QP9699-ye-50ug |
50 μg | |||||||
16048601
|
enQuireBio™
QP9699-ye-100ug |
100 μg |
£800.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16078601
|
enQuireBio™
QP9699-ye-200ug |
200 μg | |||||||
16088601
|
enQuireBio™
QP9699-ye-500ug |
500 μg |
£1930.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16068601
|
enQuireBio™
QP9699-ye-1mg |
1 mg | |||||||
Specifications
NP_001139773.1 | |
Serpin A3-2 Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
100272170 | |
Research Use Only | |
SERPINA3-2 | |
Recombinant Protein | |
Yeast | |
LPENVVVKDQ HRRVDGHTLA SSNTDFAFSL YKQLALKNPN KNVILSPLSV SIALAFLSLG ARGSTLTEIL EGLKFNLTEI QEKEIHHSFQ HLLQALNQPS NQLQLSVGNA MFVQEELKLL DKFIEDAQVL YSSEAFPTNF RDSEAARSLI NDYVKNKTQG KIEELFKYLS PRTELVLVNY IYFKAQWKTP FDPKHTEQAE FHVSDNKTVE VPMMTLDLET PYFRDEELGC TLVELTYTSN DSALFILPDE GKMRDLEAKL TPETLTRWRN SLQPRRIHEL YLPKFSIKSN YELNDILSQL GIRKIFANAD LSGITGTADL VVSQVVHGAA LDVDEEGTEG VAATGIGIER TFLRIIVRVN RPFLIAVVLK DTQSIIFLGK VTNPSEA | |
Greater than 90% as determined by SDS-PAGE. |