missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type B Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type B, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Name | Botulinum neurotoxin type B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | botB |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078421
|
enQuireBio™
QP9517-YE-100UG |
100 μg |
£800.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018431
|
enQuireBio™
QP9517-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088421
|
enQuireBio™
QP9517-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008431
|
enQuireBio™
QP9517-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028431
|
enQuireBio™
QP9517-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098421
|
enQuireBio™
QP9517-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Botulinum neurotoxin type B Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
botB | |
Recombinant Protein | |
Yeast | |
MPVTINNFNY NDPIDNNNII MMEPPFARGT GRYYKAFKIT DRIWIIPERY TFGYKPEDFN KSSGIFNRDV CEYYDPDYLN TNDKKNIFLQ TMIKLFNRIK SKPLGEKLLE MIINGIPYLG DRRVPLEEFN TNIASVTVNK LISNPGEVER KKGIFANLII FGPGPVLNEN ETIDIGIQNH FASREGFGGI MQMKFCPEYV SVFNNVQENK GASIFNRRGY FSDPALILMH ELIHVLHGLY GIKVDDLPIV PNEKKFFMQS TDAIQAEELY TFGGQDPSII TPSTDKSIYD KVLQNFRGIV DRLNKVLVCI SDPNININIY KNKFKDKYKF VEDSEGKYSI DVESFDKLYK SLMFGFTETN IAENYKIKTR ASYFSDSLPP VKIKNLLDNE IYTIEEGFNI SDKDMEKEYR GQNKAINKQA YEEISKE | |
Greater than 90% as determined by SDS-PAGE. |