missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant C. difficile Toxin B/TcdB Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 6246-GT-020

319.65 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18390408

  • £418.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to hydrolyze UDP-Glucose. The specific activity is >45 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P18177
Supplied as a 0.2 μm filtered solution in Tris and NaCl.
M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 64 kDa
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
toxB
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Enzyme Activity
4914074
Toxin B/TcdB
E. coli-derived c. difficile Toxin B/TcdB protein Met1-Leu543, with a C-terminal 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
tcdB
Recombinant
Toxin B/TcdB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers