Learn More
R&D Systems™ Recombinant Canine CCL4/MIP-1 beta Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 5839-MB-025/CF
 View more versions of this product
Product Code. 18196380
  • £327.21 / 25µg
Explore more special offers
Description
Description
The Recombinant Canine CCL4/MIP-1 beta Protein is derived from E. coli. The Recombinant Canine CCL4/MIP-1 beta Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
CCL4/MIP-1 beta
E. coli-derived canine CCL4/MIP-1 beta protein Ala24-Asn92
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7.8 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Product Suggestions